Learn More
Strem, An Ascensus Company CAS# 210169-40-7. (S)-(+)-5,5'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole, min. 98% (S)-(+)-DTBM-SEGPHOS MFCD09753003. 5G
SDP
Supplier:  Strem, An Ascensus Company 1500675G
Encompass
CAS# 210169-40-7. 5G. (S)-(+)-5,5'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole, min. 98% (S)-(+)-DTBM-SEGPHOS. MFCD09753003. Molecular Formula: C74H100O8P2 Molecular Weight: 1179.53. Color/Form: off-white pwdr. Strem#: 15-0067 www.strem.com/catalog/v3/15-0067/
Catalog No. 50-195-9583