Learn More
Medchemexpress LLC DTBT 500mg | 500MG
SDP
Supplier:  Medchemexpress LLC HY114856500MG
Encompass_Preferred
DTBT is a six-membered heterocyclic ring that is commonly used in the preparation of organic electronic devices such as P-type organic semiconductors (OFETs) and P-type organic photodiodes (OLEDs) DTBT has good performance in electron transmission and photoelectric conversion DTBT polymerizes to form donor-acceptor polymers which are used to prepare solar cells[1]
Catalog No. 50-002-29517