Learn More
GA International DTermo™ – DYMO-Compatible Paper Labels
Supplier:  GA International EDY040RE
Encompass_Preferred
Paper DYMO-compatible labels for use in the lab or office. These general use labels are perfect for improving laboratory organization, labeling file folders, general labware, notebooks, boxes, and shipping containers. Not suitable for deep-freeze applications.
Catalog No. NC1397726
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.