Learn More
Cayman Chemical DthreoPDMP hydrochloride
SDP
Supplier:  Cayman Chemical 1017810 MG
Encompass_Preferred
A glucosylceramide synthase inhibitor; inhibits glucosylceramide synthase by 50% at a 5 µM in an enzyme assay; the active component of racemic DL-threo-PDMP; inhibits the synthesis of glucosylceramide synthase and lactosylceramide in B16 melanoma cells at 25 µM; inhibits B4GALT6 and prevents lactosylceramide synthesis; inhibits ganglioside biosynthesis, reduces long-term potentiation (LTP) in mouse hippocampal CA1 neurons, and impairs learning in the four-pellet taking test in mice
Catalog No. 50-288-9113