Learn More
Medchemexpress LLC DTPA-tetra (t-Bu ester) | 174267-71-1 | 99.0% | C30H55N3O10 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYA0074
Encompass_Preferred
DTPA-tetra (t-Bu ester) is a bifunctional chelator (BFC) and a macrocyclic DPTA derivative used for tumor pre-targeting. It can be used for the conjugation of peptides and radionuclides, serving as a laboratory chemical for the manufacture of substances.
  • Bifunctional chelator (BFC)
  • Macrocyclic DPTA derivative
  • Used for tumor pre-targeting
  • Used for conjugation of peptides and radionuclides
  • Suitable for use as a laboratory chemical
  • Utilized in the manufacture of substances
Catalog No. 50-003-58405