Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DTPD-Q | 252950-56-4 | 99.7% | 318.37 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY163650100MG

Encompass_Preferred

DTPD-Q is the transformation product (TATPs) of the tire additive N,N′-diphenyl-p-phenylenediamine (DTPD). It accumulates and raises ecological risk in the environment.

  • For research use only.
  • Appearance: Solid, yellow to brown.
  • Solubility: DMSO : 2 mg/mL (6.28 mM; ultrasonic and warming and heat to 60°C; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO).
  • Storage: Powder -20°C 3 years, 4°C 2 years. In solvent -80°C 6 months, -20°C 1 month.

Catalog No. 50-003-92973


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.