Learn More
Medchemexpress LLC DTPD-Q | 252950-56-4 | 99.7% | 318.37 | C20H18N2O2 | 10 MG

Supplier: Medchemexpress LLC HY16365010MG
DTPD-Q is a quinone transformation product of the tire antioxidant N,N'-diphenyl-p-phenylenediamine (DTPD), supplied as a solid research chemical for environmental and toxicological studies. Products are intended for research use only and are not for human or clinical use.
- Quinone transformation product of N,N'-diphenyl-p-phenylenediamine (DTPD).
- High purity: 99.68%.
- Molecular formula C20H18N2O2; molecular weight 318.37.
- Solid powder form, packaged in milligram quantities.
- Soluble in DMSO at 2 mg/mL with ultrasonic warming; heat to 60°C to improve solubility.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.