Learn More
Abcam DTSSP crosslinker, Lysine selective crosslinker, 50MG
Supplier: Abcam AB14561650MG
MW 608.51 g/mol, Purity >80%. Membrane-impermeable lysine selective crosslinker. Induces cell-surface crosslinking without interfering with intercellular proteins. Facilitates the structural characterization of proteins with mass spectrometry. Thiol cleavable.
The product is subject to the following: Abcam Restricted Use Statement