Learn More
Selleck Chemical LLC DTT
SDP
Supplier:  Selleck Chemical LLC S68291G
Encompass_Preferred
DTT (Dithiothreitol) a small-molecule redox reagent is a standard agent for reducing disulfide bonds between and within biological molecules Solutions are best fresh-prepared
Catalog No. 50-000-02528