Learn More
Lascar Electronics Inc Wireless Dual Channel Thermocouple Data Logger
SDP
Supplier:  Lascar Electronics Inc ELMOTEDTC
Encompass_Preferred
The EL-MOTE-DTC is a dual channel, Cloud-enabled, wireless data logger, designed to monitor temperature in a range of indoor & outdoor applications. EasyLog Cloud harnesses the power of IoT to automate data logging & alert notifications - perfect for compact systems with just a few sensor requirements to corporate solutions with thousands of devices around the globe. -Alarm functions include a built in sounder & LED-Quick, wireless App based configuration-Stores data internally if device loses WiFi connection & automatically uploads it to the Cloud once reconnected-Suitable for indoor & outdoor applications-Industry leading 2 years battery life-Add first logger for free Manage multiple data loggers in locations around the world & view them using your Internet Browser or EasyLog Cloud App. The EasyLog Cloud offers unlimited & secure data storage, advanced alarm notifications to multiple users & powerful analysis & reporting from all your devices.
Catalog No. NC1856763