Learn More
Grainger Dual Foot Lock-On Chuck 14 long
Supplier:  Grainger 140X06
Encompass_Preferred
The Haltec Dual Foot Air Chuck is a pneumatic tool accessory used to inflate tires. It has a 1/4 inch NPT inlet and dual chucks that allow simultaneous inflation of two tires. The chucks have a 360 degree swivel inlet ball for easy maneuvering. The chucks are made of zinc plated steel with rubber dust caps. The maximum working pressure is 160 PSI. This air chuck allows efficient inflation of multiple tires.
Catalog No. 50-275-0770