Learn More
Electron Microscopy Sciences Dual Geological Thin Section Holder, 9.5 mm Pin
SDP
Supplier:  Electron Microscopy Sciences 7594118
Encompass
This thin section holder made from aluminum with copper clips and brass screws. The unit measures 55x58x8 mm and it holds two standard geological thin sections or slides of 47 x 27 mm. The holder has convenient tweezer inserts which allow for easy loading. Clips are provided for secure holding.
Type: 9.5 mm Pin
Catalog No. 50-190-1645