Learn More
Trajan Scientific PEEK Tubing, Dual Layer Color Coded PEEK Tubing (Orange) - 1/16" OD x .020" ID x 100FT, 5000 PSI, 345 BAR
Supplier:  Trajan Scientific VAL.JRTD6010100
Encompass_Preferred
PEEK Tubing, Dual Layer Color Coded PEEK Tubing (Orange) - 1/16" OD x .020" ID x 100FT, 5000 PSI, 345 BAR. Dual Layer PEEK tubing eliminates any concern that a critical sample stream could be contaminated by pigments used to color code the tubing. It looks like any other color-coded tubing at first glance, but a closer look reveals that the pigmented layer surrounds a separate but integrally-bonded inner layer of natural PEEK.
Catalog No. 50-138-5090