Learn More
Thermco Products Inc DUAL PROBE Min/Max Time Date Stamp Digital Alarm Thermometer - Monitor Freezer and Refrigerator simultaneously. 5ml Vaccine Bottle probes filled with Bio-Safe medium. Audible & Visual alarms. -50 to 70C x 0.1C, Factory Certified @ -20C & 4C
Supplier:  Thermco Products Inc ACC8100MATB
Encompass_Preferred
This product was recently added by customer request, and is available for your convenience. We strive to provide our customers with a one-stop shop for the entire scientific supplies category. More relevant content may be added as customer demand increases.
Catalog No. NC0755297