Learn More
Analytical Sales & Services Dual Reservoir with Lid, 25/pk
Supplier:  Analytical Sales & Services 55001
Encompass
Polypropylene Dual Reservoir with Lid. One 50mL well on one side. Twelve 5mL wells on opposite side.
Catalog No. NC1556664