Learn More
LifeSensors DUB Cocktail 25µg
SDP
Supplier:  LifeSensors DB05990025
Encompass_Preferred
This DUB cocktail contains an optimized combination of DUBs to be used as part of the UbiTest assay. Prior experiments with single broad-spectrum DUBs did not completely cleave all ubiquitin chains. This cocktail was designed for complete DUB digestion as part of any assay.
Catalog No. NC2040391