Learn More
Lascar Electronics Inc Wireless High Accuracy Temperature Data Logger with Dual Thermistor Probes
SDP
Supplier:  Lascar Electronics Inc ELMOTEDTP+
Encompass_Preferred
The EL-MOTE-DTP+ is a Cloud-enabled, high accuracy, wireless data logger, designed to monitor temperature in a large range of indoor & outdoor applications. EasyLog Cloud harnesses the power of IoT to automate data logging & alert notifications - perfect for both compact systems with just a few sensor requirements to corporate solutions with thousands of devices around the globe. Alarm functions include a built in sounder & LEDQuick, wireless App based configStores data internally if device loses WiFi connection & automatically uploads it to the Cloud once reconnectedSuitable for indoor & outdoor applications2 years battery lifeAdd first logger for freeManage multiple data loggers in locations around the world & view them using your Internet Browser or EasyLog Cloud App. The EasyLog Cloud offers unlimited & secure data storage, advanced alarm notifications to multiple users & powerful analysis & reporting from all your devices.
Catalog No. NC1856677