Learn More
ATCC Dulbecco's Modified Eagle's Medium (DMEM)
Supplier:  ATCC 302002
Encompass
Dulbecco's Modified Eagle's Medium (DMEM) is modified to contain 4 mM L-glutamine, 4500 mg/L glucose, 1 mM sodium pyruvate, and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-4494
 Restricted Item