Learn More
Selleck Chemical LLC Duloxetine S5071-25mg
SDP
Supplier:  Selleck Chemical LLC S507125MG
Encompass_Preferred
Duloxetine (LY-248686) is a potent inhibitor of serotonin (5-hydroxytryptamine 5-HT) and noradrenaline (NE) uptake in vitro and in vivo and is 3- to 5-times more effective at inhibiting 5-HT uptake
Catalog No. 50-311-0931