Learn More
TARGETMOL CHEMICALS INC DULOXETINE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T706150MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. Duloxetine is an inhibitor of serotonin-norepinephrine reuptake(Ki of 4.6 nM)with treatment of major depressive disorder and generalized anxiety disorder. purity: 99%
Catalog No. 50-257-6358