Learn More
Apexbio Technology LLC Duloxetine HCl 136434-34-9 100mg
SDP
Supplier:  Apexbio Technology LLC A8421100
Encompass_Preferred
Duloxetine hydrochloride (CAS 136434-34-9) is a small molecule compound classified as a selective inhibitor of serotonin and norepinephrine reuptake By blocking the reabsorption of these neurotransmitters at the synaptic cleft duloxetine modulates serotonergic and noradrenergic signaling within the central nervous system This pharmacological action has rendered it a valuable tool in research models investigating neurochemical pathways involved in mood regulation pain perception and stress response Duloxetine HCl is commonly utilized in studies of depression anxiety and chronic pain to elucidate monoaminergic mechanisms and to evaluate therapeutic strategies targeting neurotransmitter reuptake
Catalog No. 50-000-96250