Learn More
Fine Science Tools Dumont Vessel Cannulation Forceps, 11 cm
SDP
Supplier:  Fine Science Tools 1128011
Encompass
Product: Dumont Vessel Cannulation Forceps - 0.35mm OD/11cm/ Tip Shape: Angled/ Tubing Size: 0.35mm OD/ Alloy: Inox/ Length: 11cm/ Unit: Each
Catalog No. NC1160991