Learn More
Fine Science Tools Dumont #4 Forceps, Straight, 0.13 x 0.08 mm, 11 cm
SDP
Supplier:  Fine Science Tools 1124130
Encompass
Style: #4/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.13 x 0.08mm/ Alloy / Material: Dumoxel/ Length: 11cm
Catalog No. NC9091939