Learn More
Medchemexpress LLC Dup-721 | 104421-21-8 | 98.0% | 276.29 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13961850MG
Encompass_Preferred
DuP-721 is a broad-spectrum, orally active antibacterial agent effective against various clinically susceptible and resistant bacteria, especially *M. tuberculosis*. It inhibits *Mycobacterium tuberculosis* strains (1.5-4 μg/ml) and shows no cross-resistance to conventional antituberculosis drugs. In *in vivo* studies, DuP-721 demonstrates protective effects against *M. tuberculosis* infection in mice.
  • Effective against susceptible and resistant bacteria
  • Targets *M. tuberculosis*
  • Inhibits *Mycobacterium tuberculosis* (1.5-4 μg/ml)
  • No cross-resistance to antituberculosis drugs
  • Protects against *M. tuberculosis* infection in mice
  • Inactive against *M. avium* and *M. intracellulare* (>250 μg/ml)
  • Inhibits *M. gordonoe* and *M. fortuitum* (3.9 μg/ml)
  • Inhibits *M. kansassi* (1.95 μg/ml)
  • Inhibits *M. scrofulaceum* (15.6 μg/ml)
Catalog No. 50-003-49281