Learn More
TARGETMOL CHEMICALS INC DuP 747 HCl 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T25354L1MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DuP 747 HCl is a selective kappa agonist with analgesic activity.DuP 747 consists of two conformations. Purity 99.81%
Catalog No. 50-287-9751