Learn More
Medchemexpress LLC DUPA(OtBu)-OH | 1026987-94-9 | MFCD28898588 | 98.0% | 488.57 g/mol | C23H40N2O9 | 50 MG

Supplier: Medchemexpress LLC HY10359150MG
DUPA(OtBu)-OH is a chemical precursor used to introduce the DUPA targeting moiety into PSMA-targeted drug conjugates. It enables selective delivery of chemotherapeutics to PSMA-expressing prostate cancer cells and supports conjugation strategies that promote tumor-selective release. Supplied as a solid and as a DMSO solution for research applications.
- Used as a precursor for PSMA-targeted drug conjugates.
- Supports disulfide-linked conjugation for triggered drug release.
- Chemical formula C23H40N2O9.
- Molecular weight 488.57 g/mol.
- Purity 98.0% (manufacturer stated).
- Available in solid quantities and as a 10 mM solution in DMSO.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.