Learn More
Medchemexpress LLC HY-P9926 1mg Medchemexpress, Dupilumab CAS:1190264-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99261MG
Encompass_Preferred
Medchemexpress, HY-P9926 1mg Dupilumab CAS:1190264-60-8 Dupilumab (REGN-668) is a fully human mAb to IL-4 receptor α (IL-4Rα) that inhibits both IL-4 and IL-13 signaling, markedly improved moderate-to-severe atopic dermatitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1775