Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR laboratory bottles
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z30517010EA
Encompass
Reusable bottles and caps for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.
Catalog No. 50-290-1423