Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR Youtility bottle GL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7420144EA
Encompass
Specially shaped gripping zones on both sides of the bottle for easier and safer handlingOpening or closing of the bottle is faster and fully compatible with DIN GL 45 closures and other accessoriesSlimmer YOUTILITY bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigeratorsPre-defined labelling area is compatible with DURAN self-adhesive YOUTILITY bottle labelsNominal volume is shown at the top of the easy-to-read graduation scale for fast determination of the volumesSequentially sized bottles are available in four volumesMoulded from the DURAN borosilicate 3.3 pharmacopeia type 1 neutral glassRetrace Code for full product traceabilityEach bottle is supplied as a complete system with a pouring ring (cyan polypropylene) and a GL 45 cap (cyan polypropylene)
Catalog No. 50-290-6019