Learn More
MSC Norton Floor Sanding Disc, Silicon Carbide, Heavy Paper Backing, 2 in Arbor Hole
Supplier:  MSC 68835735
Encompass
These floor sanding discs are engineered for high-speed floor sanding applications, providing a quality cut and consistent performance. They are effective at blending scratches from both edger and drum sanders on a variety of floor surfaces.
  • Made with 100% silicon carbide abrasive for a quality cut
  • Easily blends scratches from edger and drum sanders
  • Heavy paper backings ensure strength and extended disc life
  • Reduces dish-out of springwood on strip and plank floors
  • Offers good value and reliable performance
  • Ideal for floor sanding and grinding applications
  • Can be used on materials such as concrete and hard woods
Catalog No. 50-283-7465