Learn More
Electron Microscopy Sciences Dust-Off Plus Refill(DPSA) 8 OZ
SDP
Supplier:  Electron Microscopy Sciences 70844
Encompass_Preferred
Contains: Formula S (Difluoroethane) - 100% safe for the ozone. It comes with a patented 360 degree Vector Valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can.
Catalog No. 50-930-494