Learn More
Medchemexpress LLC dUTP trisodium | 102814-08-4 | MFCD00084701 | 99.7% | 534.09 g·mol⁻¹ | C9H12N2Na3O14P3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW017068100MG
Encompass_Preferred
dUTP trisodium is the trisodium salt of 2'-deoxyuridine-5'-triphosphate, used in molecular biology as a uracil-containing nucleotide substitute for dTTP. It is provided as a high-purity solid and as a PCR-grade solution for incorporation into PCR and enzymatic assays; intended for research use only (not for human use).
  • High purity (>99% by HPLC) ensuring reliable incorporation.
  • Available as a solid (100 mg) and as a 100 mM PCR-grade solution.
  • Suitable for PCR and RT-PCR applications as a dTTP substitute.
  • Stable when stored per manufacturer guidance; supports long-term storage in solution at -80°C.
  • Supplied with certificate of analysis reporting assay and storage recommendations.
Catalog No. 50-003-02059