Learn More
TARGETMOL CHEMICALS INC DUVELISIB R ENANTIOMER H 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11129L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Duvelisib (R enantiomer) hydrochloride (INK1197 R enantiomer HCl) is a PI3K inhibitor. Duvelisib (R enantiomer) hydrochloride is the less active enantiomer of Duvelisib.
Catalog No. 50-258-1567