Learn More
Aviva Systems Biology Corp DUX3 Antibody - N-terminal region (ARP35847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUX3. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Human, Zebrafish
Catalog No. 50-284-476