Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS High Alloy DX Biological Tweezers Style 5, 110 mm OAL, 0.05 mm Tip width
SDP

Supplier:  Electron Microscopy Sciences 783255DX

Encompass_Preferred

The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Straight, extra fine, superior finish.
OAL: 110 mm
Tip width: 0.05 mm
Tip thickness: 0.02 mm

Catalog No. 50-193-0612


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.