Learn More
Selleck Chemical LLC Dxd E2891-100MG
SDP
Supplier:  Selleck Chemical LLC E2891100MG
Encompass_Preferred
Dxd(Exatecan derivative for ADC) is a potent DNA topoisomerase I inhibitor with an IC50 of 0 31 M
Catalog No. 50-000-39697