Learn More
Medchemexpress LLC Dxd (exatecan derivative) | 1599440-33-1 | MFCD31657444 | 99.3% | 493.48 | C26H24FN3O6 | 200 MG

Supplier: Medchemexpress LLC HY13631D200MG
Dxd (an exatecan derivative) is a potent DNA topoisomerase I inhibitor used as the cytotoxic payload in HER2-targeting antibody-drug conjugates. It has a reported in vitro IC50 of 0.31 μM, a molecular weight of 493.48, and high purity suitable for ADC synthesis and research applications.
- Potent topoisomerase I inhibitor (IC50 0.31 μM).
- High purity: 99.3%.
- Molecular formula: C26H24FN3O6.
- Molecular weight: 493.48 g/mol.
- CAS number: 1599440-33-1.
- Typical pack size: 200 mg.
- Intended for use as an ADC payload in research and conjugation workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.