Learn More
Medchemexpress LLC DYB-03 | 2982792-85-6 | 99.5% | 479.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY161313
Encompass_Preferred
DYB-03 is an orally active HIF-1α/EZH2 inhibitor. It inhibits migration, invasion, and angiogenesis of lung cancer cells and HUVECs both in vitro and in vivo. Additionally, DYB-03 induces apoptosis in A549 and H460 cells that are resistant to 2-ME2 and GSK126.
  • Is an oral active HIF-1α/EZH2 inhibitor.
  • Inhibits migration of lung cancer cells and HUVECs in vitro and in vivo.
  • Inhibits invasion of lung cancer cells and HUVECs in vitro and in vivo.
  • Inhibits angiogenesis of lung cancer cells and HUVECs in vitro and in vivo.
  • Induces apoptosis in 2-ME2- and GSK126-resistant A549 and H460 cells.
Catalog No. 50-003-94723