Learn More
Selleck Chemical LLC Dyclonine HCl S2041-50mg
SDP
Supplier:  Selleck Chemical LLC S204150MG
Encompass_Preferred
Dyclonine HCl(Dyclocaine HCl) is a hydrochloride salt form of dyclonine which is an oral anaesthetic reversibly binds to activated sodium channels on the neuronal membrane thereby decreasing the neuronal membrane s permeability to sodium ions leading to an increased threshold for excitation
Catalog No. 50-310-9689