Learn More
Medchemexpress LLC HY-B0257A 5mg , Dydrogesterone CAS:152-62-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0257A5MG
Encompass_Preferred
HY-B0257A 5mg Dydrogesterone CAS: 152-62-5 Dydrogesterone is a potent, orally active progestogen indicated in a wide variety of gynaecological conditions related to progesterone deficiency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1922