Learn More
Ethos Biosciences Tissue Marking Dye, Royal Blue, 2oz, Ethos
Supplier:  Ethos Biosciences 756302
Encompass_Preferred
Tissue Marking Dye is designed for clear and durable identification of surgical margins and specimen orientation during grossing and processing. The fast-drying formula adheres to fresh and fixed tissue surfaces, providing distinct, high-contrast color that remains visible throughout routine processing and staining. Compatible with paraffin embedding and standard histology workflows, it supports accurate specimen tracking and orientation. Available in multiple colors for precise margin designation, the dye is suitable for clinical, research, and teaching laboratories and is intended for professional laboratory use.
Catalog No. 50-005-74247