Learn More
Biotium Dyes For Labeling Carbonyls and Carbohydrates Fluorescence Microscopy Cell and Organelle Stains Cell Surface and Membrane Dyes 1/EA
Supplier:  Biotium 30143T
Encompass_Preferred
Label live cell surface glycoproteins with biotin for fixable plasma membrane staining or protein extraction GlycoLiner uses novel reactive chemistry for faster more efficient and gentler labeling compared to conventional methods Label 100 wells (200 uL) or two T75 flasks (10 mL)
Catalog No. 50-005-01420