Learn More
Cayman Chemical DYKDDDDK PeptIDtrIfluoROC 25mg
SDP
Supplier:  Cayman Chemical 3319825 MG
Encompass_Preferred
A octapeptide fusion tag; has commonly been used as an epitope tag on recombinant proteins to facilitate their detection and purification by DYKDDDDK-specific antibodies in vitro and in vivo
Catalog No. 50-294-7927