Learn More
Abcam Dylight 350 Conjugation Kit (Fast) - Lightning-Link, 3 x 10UG
Supplier:  Abcam AB2017973X10UG
Encompass_Preferred
Labeling in < 20 mins with 30 secs hands-on time using Dylight 350 Conjugation Kit (Fast) - Lightning-Link ab201797. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9150