Learn More
Vector Laboratories DyLight 649 Streptavidin 1 mg
SDP
Supplier:  Vector Laboratories SA56491
Encompass
Vector Laboratories fluorochrome-conjugated streptavidin and avidin reagents are highly purified and possess very low non-specific binding properties. They have a high affinity for biotin and can be used to detect biotinylated secondary antibodies and other macromolecules in applications such as immunofluorescence, in situ hybridization, or flow cytometry. Fluorescent signals can be easily amplified using biotinylated secondary antibodies followed by fluorochrome-labeled streptavidin or avidin. DyLight-conjugated streptavidin offers several advantages including photostability, pH independence, and brighter fluorescence. These conjugates are useful in a variety of applications, including cell and tissue-based immunofluorescent detection.
Catalog No. NC0361181