Learn More
GA International DYMO-Compatible Cryo Labels for Frozen Surfaces
Supplier:  GA International EDCS040WH
Encompass_Preferred
Dymo-Compatible CryoSTUCK - Cryogenic Labels for Frozen Surfaces, 0.5" x 1" / 12.7mm x 25.4mm, White, 2 Labels Across, 1000 L/R
Catalog No. NC1985255