Learn More
Enterprise Technology Solutions Dymo DYMO D1 Electronic Tape, 3/8"x23' Size, Black/White
SDP
Supplier:  Enterprise Technology Solutions DYM41913
Encompass_Preferred
Durable, polyester-coated D1 tape is both water-resistant and sunlight-resistant and endures temperatures from zero to 194 degrees Fahrenheit. Patented D1 tape cartridge contains easy-peel, split-back tape. Special backing is split down the middle to make separation of tape backing a snap for fast and easy label application. Tape is designed for use with Dymo 1000, 2000, 3500, 4500, 5500, LM100 Plus, LM120P, LM150, LM155, LM160, LM200, LM210D, LM220P, LM260P, LM350, LM360D, LM400, LM420P, LM450D, LM500TS, LMPCII, LMPnP, LP100, LP150, LP200, LP250, LP300, LP350, and LabelWriter Duo.
Catalog No. 50-007-40654