Learn More
Medchemexpress LLC Dynamin IN-1 | 1345853-50-0 | C23H24N2O | 50 MG

Supplier: Medchemexpress LLC HY15221550MG
Dynamin IN-1 is a potent dynamin inhibitor with an IC50 value of 1.0 μM. This compound is intended for research use only and is not sold for patient use.
- Potent dynamin inhibitor
- IC50 value of 1.0 μM
- For research use only
- Purity of 99.89%
- Molecular weight: 344.45
- Appearance: Solid, white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.