Learn More
Medchemexpress LLC Dynamin inhibitory peptide | 251634-21-6 | 96.1% | 1121.25 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP10831MG
Encompass_Preferred
Dynamin inhibitory peptide competitively blocks the binding of dynamin to amphiphysin, thereby preventing endocytosis. It also acts to block the dopamine D3 effect on GABAA receptors. This product is intended for research use only and is not sold to patients.
  • Competitively blocks binding of dynamin to amphiphysin.
  • Prevents endocytosis.
  • Blocks the dopamine D3 effect on GABAA receptors.
Catalog No. 50-004-22201