Learn More
ALL PRINTING RESOURCE Dyne Testing Pen Kit, - Levels 30-60 (even numbered)
Supplier:  ALL PRINTING RESOURCE DTPKIT 3060
Encompass
This item has a minimum order quantity of 2 per supplier requirements. 16 Standard levels from 30 through 60 dynes/cm. Specifically designed for fast, accurate on-line use by printers, coaters, and laminators, also ideal for process control or field use by suppliers of film, sheet, coated board, inks, coatings, and adhesives. Spring loaded tip meters fluid feed and guards against contamination. Reliable enough for many R & D and lab applications. Hundreds of tests from each marker. Usable on virtually all smooth non-absorptive substrates.
Catalog No. NC9868418